Please login to your online account to display your discounted pricing

MRS 2179 tetrasodium salt, Tocris


Selective P2Y1 antagonist

Quantity 50mg

Manufacturer: r & d systems  090050

 View more versions of this product

Catalog No. 09-005-0

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 513.16
Chemical Name or Material MRS 2179 tetrasodium salt
Assay Percent Range >98%
Molecular Formula C11H13N5O9P2Na4
CAS 1454889-37-2
Synonym 2'-Deoxy-N6-methyladenosine 3', 5'-bisphosphate tetrasodium salt
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material MRS 2179 tetrasodium salt
Assay Percent Range >98%
Molecular Formula C11H13N5O9P2Na4
Formula Weight 513.16
Synonym 2'-Deoxy-N6-methyladenosine 3', 5'-bisphosphate tetrasodium salt
Accessories

Desiccate at -20°C