Please login to your online account to display your discounted pricing

MRS 1845, Tocris

Potent SOC inhibitor; blocks capacitative Ca2+ entry

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material MRS 1845
Assay Percent Range >98%
Molecular Formula C21H22N2O6
Formula Weight 398.41
Synonym 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-1-(2-propynyl)-3,5-pyridinedicarboxylic acid ethyl, methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

186610

 
r & d systems
RDS186610
10mg Each for $221.01

186650

 
r & d systems
RDS186650
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 398.41
Quantity 10mg
Chemical Name or Material MRS 1845
CAS 544478-19-5
Synonym 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-1-(2-propynyl)-3,5-pyridinedicarboxylic acid ethyl, methyl ester
Recommended Storage Store at +4°C
Assay Percent Range >98%
Molecular Formula C21H22N2O6
Safety and Handling

Store at +4°C