Please login to your online account to display your discounted pricing

MRS 1706, Tocris

Potent and selective A2B inverse agonist

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material MRS 1706
Assay Percent Range >96%
Molecular Formula C27H29N5O5
Formula Weight 503.56
Synonym N-(4-Acetylphenyl)-2-[4-(2,3,6,7-tetrahydro-2,6-dioxo-1,3-dipropyl-1H-purin-8-yl)phenoxy]acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

158410

 
r & d systems
158410
10mg Each for $225.77

158450

 
r & d systems
158450
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 503.56
Quantity 10mg
Chemical Name or Material MRS 1706
Assay Percent Range >96%
Molecular Formula C27H29N5O5
CAS 264622-53-9
Synonym N-(4-Acetylphenyl)-2-[4-(2,3,6,7-tetrahydro-2,6-dioxo-1,3-dipropyl-1H-purin-8-yl)phenoxy]acetamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C