MRO Plus Heavy Duty Absorbent Mat Roll
Avoid slips, trips, and falls with this great addition to your absorbent program. The Brady™ MRO Plus Heavy Rolls absorbs all industrial liquids (water, petroleum-based and chemical-based fluids). Perforated for easy tears.
$99.76 - $108.90
Specifications
Quantity 1 Bale/ 2 Rolls
Absorbancy 14.5 gl/gs
For Use With (Application) Chemical resistant sorbent for water, oil and solvent base liquids
Material Polypropylene
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Capacity Dimensions (L x W) Price Quantity  
19-103-111 Brady™
BRO152
38 gal. 150 ft. x 15 in. Pack of 2 Each for $99.76
19-103-112 Brady™
BRU152
38 gal. 150 ft. x 15 in. Pack of 2 Each for $108.90
Specifications
Description
  • Absorbs all industrial liquids (water, petroleum-based and chemical-based fluids)
  • 3-ply construction makes it stronger, more durable and reduces lint
  • Gray color masks drips and spills
  • Perforated every 12 in. so you can take only what is needed and reduce waste 
Description & Specifications
Specifications
1 Bale/ 2 Rolls
14.5 gl/gs
Chemical resistant sorbent for water, oil and solvent base liquids
Polypropylene