Please login to your online account to display your discounted pricing

MRK 560, Tocris™

γ-secretase inhibitor; attenuates amyloid plaque deposition

$270.88 - $1,140.36

Chemical Identifiers

Chemical Name or Material MRK 560
Assay Percent Range >99%
Molecular Formula C19H17ClF5NO4S2
Formula Weight 517.92
Synonym N-[cis-4-[(4-Chlorophenyl)sulfonyl]-4-(2,5-difluorophenyl)cyclohexyl]-1,1,1-trifluoromethanesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

400050

 
r & d systems
RDS400050
50mg Each for $1,140.36

400010

 
r & d systems
RDS400010
10mg Each for $270.88
Description & Specifications

Specifications

Formula Weight 517.92
Quantity 50mg
Chemical Name or Material MRK 560
Assay Percent Range >99%
Molecular Formula C19H17ClF5NO4S2
Synonym N-[cis-4-[(4-Chlorophenyl)sulfonyl]-4-(2,5-difluorophenyl)cyclohexyl]-1,1,1-trifluoromethanesulfonamide
CAS 677772-84-8
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C