Please login to your online account to display your discounted pricing

MR 16728 hydrochloride, Tocris

Stimulates ACh release

$195.49 - $816.90

Chemical Identifiers

Chemical Name or Material MR 16728 hydrochloride
Assay Percent Range >98%
Molecular Formula C23H36N2O·HCl
Formula Weight 393.01
Synonym N-[3-(1-Hexahydroazapinyl)propyl]-α-cyclohexyl-benzeneacetamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

053750

 
r & d systems
053750
50mg Each for $816.90

053710

 
r & d systems
053710
10mg Each for $195.49
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 393.01
Assay Percent Range >98%
Recommended Storage Desiccate at -20°C
Synonym N-[3-(1-Hexahydroazapinyl)propyl]-α-cyclohexyl-benzeneacetamide hydrochloride
Molecular Formula C23H36N2O·HCl
Chemical Name or Material MR 16728 hydrochloride
CAS 207403-36-9
Safety and Handling

Desiccate at -20°C