Please login to your online account to display your discounted pricing

MR 16728 hydrochloride, Tocris


Stimulates ACh release

Quantity 50mg

Manufacturer: r & d systems  053750

 View more versions of this product

Catalog No. 053750

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material MR 16728 hydrochloride
Quantity 50mg
CAS 207403-36-9
Recommended Storage Desiccate at -20°C
Assay Percent Range >98%
Molecular Formula C23H36N2O·HCl
Formula Weight 393.01
Synonym N-[3-(1-Hexahydroazapinyl)propyl]-α-cyclohexyl-benzeneacetamide hydrochloride

Specifications

Chemical Name or Material MR 16728 hydrochloride
Assay Percent Range >98%
Molecular Formula C23H36N2O·HCl
Formula Weight 393.01
Synonym N-[3-(1-Hexahydroazapinyl)propyl]-α-cyclohexyl-benzeneacetamide hydrochloride
Accessories

Desiccate at -20°C