missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[Mpr1, Val4, D-Arg8] Vasopressin, Alfa Aesar™

For cellular and molecular biology applications

$89.20 - $151.55

Specifications

Product Type [Mpr1, Val4, D-Arg8] Vasopressin
Molecular Weight 1040.20
Storage Requirements -30° to -10°C
Sequence Mpr-Tyr-Phe-Val-Asn-Cys-Pro-D-Arg-Gly-NH2 (Disulfide bridge: Mpr1-Cys6)
Format Solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66210EXD
View Documents
Thermo Fisher Scientific
J66210EXD
0.5mg Each for $89.20
 
AAJ66210MCR
View Documents
Thermo Fisher Scientific
J66210MCR
1mg Each for $151.55
 
Specifications

Specifications

[Mpr1, Val4, D-Arg8] Vasopressin
-30° to -10°C
Solid
RUO
1040.20
Mpr-Tyr-Phe-Val-Asn-Cys-Pro-D-Arg-Gly-NH2 (Disulfide bridge: Mpr1-Cys6)
Deamino-Cys-Tyr-Phe-Val-Asn-Cys-Pro-D-Arg-Gly-NH2
HPLC

For Research Use Only