Please login to your online account to display your discounted pricing

MPPG, Tocris

Group III/group II mGlu antagonist; More selective for group III than group II

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material MPPG
Assay Percent Range >98%
Molecular Formula C9H12NO5P
Formula Weight 245.17
Synonym (RS)-α-Methyl-4-phosphonophenylglycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

085350

 
r & d systems
RDS085350
50mg Each for $976.15

085310

 
r & d systems
RDS085310
10mg Each for $239.63
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 245.17
Chemical Name or Material MPPG
Assay Percent Range >98%
Molecular Formula C9H12NO5P
CAS 169209-65-8
Synonym (RS)-α-Methyl-4-phosphonophenylglycine
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature