missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

VAI MP Knee-High Boots

MP Knee-High Boots feature elastic tops and ankles and are individually packaged in pairs of two.

$933.83 - $933.83

Specifications

Material Microporous Fabric
Color White
Disposable Single-use
Closure Type Elastic with Ties
Sole Type Non-skid
View More Specs

Products 3
Catalog Number Mfr. No. Size Description Price Quantity  
Catalog Number Mfr. No. Size Description Price Quantity  
19546554
View Documents Promotion Details
VAI
1700PMCNSS17039
Small/Medium 10 in. Opening Size
Case of 100 Pair for $933.83
 
19546555
View Documents Promotion Details
VAI
1700PMCNSS17040
Large/X-Large 12 in. Opening Size
Case of 100 Pair for $933.83
 
19546556
View Documents Promotion Details
VAI
1700PMCNSS17041
2X-Large/3X-Large 14 in. Opening Size
Case of 100 Pair for $933.83
 
Description

Description

  • Heavy weight, non-woven and high quality microporous (MP) coated material
  • Suited for use where maximum protection to product or wearer is required
  • Excellent liquid resistance, especially to chemicals
Specifications

Specifications

Microporous Fabric
Single-use
Non-skid
Individually Packaged Pairs
457 mm
White
Elastic with Ties
100/Cs.
18 in.
Controlled Environments Boot Covers
SDS
missing translation for 'documents'

missing translation for 'documents'