missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ YNB, Powdered Media: With Ammonium Sulfate
Can be prepared by adding a carbon source, the appropriate amino acids, and agar if necessary
Supplier: MP Biomedicals™ 114027522
Description
- Nitrogen-based yeast
Yeast cell culture, Gene expression, Transcription regulation
Specifications
15°C to 30°C | |
Microbiology Media | |
For vigorous growth of Saccharomyces cerevisiae |
Powder | |
Nitrogen Source: Ammonium Sulfate 5000mg/L, Vitamins: Biotin 0.002mg/L, Calcium Pantothenate 0.4mg/L, Folic Acid 0.002mg/L, Inositol 2mg/L, Niacin 0.4mg/L, P-Aminobenzoic Acid 0.2mg/L, Pyridoxine Hydrochloride 0.4mg/L, Riboflavin 0.2mg/L, Thiamine Hydrochloride 0.4mg; Trace Elements: Boric Acid 0.5mg, Copper Sulfate 0.04mg, Potassium Iodide 0.1mg, Ferric Chloride 0.2mg, Manganese Sulfate 0.4mg, Sodium Molybdate 0.2mg, Zinc Sulfate 0.4mg; Salts: Magnesium Sulfate 500mg, Sodium Chloride 100 mg, Potassium Phosphate monobasic 1000mg, Calcium Chloride 100mg | |
454 g |