Please login to your online account to display your discounted pricing

MP Biomedicals Yeast Lytic Enzyme

From Arthrobacter luteus

$79.95 - $1,035.41

Specifications

Color Beige
Concentration ≥100000U/g
Product Name Yeast Lytic Enzyme
Format Lyophilized Powder
For Use With (Application) Cell lysis, spheroplasting and Glucan hydrolysis
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19012310

 
mp biomedicals
19012310
10U Each for $585.70

ICN19012301

 
mp biomedicals
19012301
1U Each for $79.95

ICN19012325

 
mp biomedicals
19012325
25U Each for $1,035.41
Description & Specifications

Specifications

Color Beige
Concentration ≥100000U/g
Product Name Yeast Lytic Enzyme
Format Lyophilized Powder
For Use With (Application) Cell lysis, spheroplasting and Glucan hydrolysis
pH 5 to 10
Source Arthrobacter luteus
Storage Requirements 0°C

Activity: 100,000U/g. Unit Definition: One unit will produce a Δ A800 of 0.001 per minute at pH 7.5 and 25°C, using a suspension of brewers yeast as a substrate in a 3mL reaction mixture. This enzyme catalyzes the lysis of yeast cells and hydrolyzes glucose polymers with β-1,3-linkages (e.g., glucan). It is also useful for preparing yeast spheroplasts. Lyophilized powder.