Please login to your online account to display your discounted pricing

MP Biomedicals Yeast Lytic Enzyme


From Arthrobacter luteus

Manufacturer: mp biomedicals  19012325

 View more versions of this product

Catalog No. ICN19012325

  • / Each
 

Description & Specifications

Specifications

Source Arthrobacter luteus
pH 5 to 10
Concentration ≥100000U/g
Product Name Yeast Lytic Enzyme
Quantity 25U
For Use With (Application) Cell lysis, spheroplasting and Glucan hydrolysis
Storage Requirements 0°C
Format Lyophilized Powder
Concentration ≥100000U/g
Quantity 25U
Color Beige
Format Lyophilized Powder