Please login to your online account to display your discounted pricing

MP Biomedicals Yeast Lytic Enzyme


From Arthrobacter luteus

Manufacturer: mp biomedicals  19012310

 View more versions of this product

Catalog No. ICN19012310

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 0°C
Quantity 10U
Product Name Yeast Lytic Enzyme
Format Lyophilized Powder
pH 5 to 10
Color Beige
Source Arthrobacter luteus
Format Lyophilized Powder
Concentration ≥100000U/g
For Use With (Application) Cell lysis, spheroplasting and Glucan hydrolysis
Concentration ≥100000U/g
Quantity 10U