Please login to your online account to display your discounted pricing

MP Biomedicals Yeast Lytic Enzyme


From Arthrobacter luteus

Manufacturer: mp biomedicals  19012301

 View more versions of this product

Catalog No. ICN19012301

  • / Each
 

Description & Specifications

Specifications

pH 5 to 10
Color Beige
For Use With (Application) Cell lysis, spheroplasting and Glucan hydrolysis
Concentration ≥100000U/g
Product Name Yeast Lytic Enzyme
Storage Requirements 0°C
Source Arthrobacter luteus
Format Lyophilized Powder
Quantity 1U