Please login to your online account to display your discounted pricing

MP Biomedicals Trypsin, 1 - 300, From Hog Pancreas

From Hog Pancreas

Manufacturer: mp biomedicals  10314080

 View all options for this product

Catalog No. ICN10314080


Description & Specifications


Format Powder
Molecular Weight 24000
Name Trypsin
pH Range ∽8
Quantity 100g
Source Hog
Storage Requirements 4°C

Trypsin, 1 - 300, [9002-07-7]


Dry Powder