Please login to your online account to display your discounted pricing

MP Biomedicals Trypsin, 1 - 300, From Hog Pancreas

From Hog Pancreas

$187.27 - $502.10

Specifications

Format Powder
Name Trypsin
pH Range ∽8
Source Hog
Storage Requirements 4°C
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10314025

 
mp biomedicals
10314025
25g Each for $187.27

ICN10314080

 
mp biomedicals
10314080
100g Each for $502.10
Description & Specifications

Specifications

Format Powder
Name Trypsin
pH Range ∽8
Source Hog
Storage Requirements 4°C

Trypsin, 1 - 300, [9002-07-7]

Parenzymol

Dry Powder