missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Superbroth: Regular

Molecular biology certified bacterial growth medium for the growth of E. coli

Manufacturer:  MP Biomedicals™ 113010041

 View more versions of this product

Catalog No. MP113010041



Specifications

Specifications

Molecular Biology Certified™
Capsule
5000g
Super Broth
Molecular biology
Tryptone-B 35g/L, Yeast extract 20g/L, NaCI 5g/L
15° to 30°C
For obtaining high yields of lambda phage and phagemids in Escherichia coli
Documents