missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Soy Protein Isolated
Protein 92%
Manufacturer: MP Biomedicals™ 0290545605
Description
A source of protein used in preparing animal research diets.
Specifications
Protein (N x 6.25) 92%, Moisture 6%, Ash 4.1%, Fat (PE extract) 0.8%, Fiber (crude) 0.25%, Calcium 0.15%, Phosphorus 0.8%, Sodium 1.3%, Potassium 0.05%, Particle Size 90% through 100 mesh, pH (water slurry) 7.1, Standard Plate Count <10000/g, Salmonella Negative, Yeast-Mold <100/g, Trypsin Inhibitor 4.9 to 7.3mg/g protein | |
Powder | |
2260 g | |
Soy Protein Isolated |
Soy Protein isolated used in preparing animal research diets | |
Pellet | |
15°C to 30°C |