Please login to your online account to display your discounted pricing

MP Biomedicals Ribonuclease From Bovine Pancreas

Manufacturer: mp biomedicals  0210107505

 View all options for this product

Catalog No. ICN10107505


Description & Specifications


Product Name Ribonuclease
Format Powder
Concentration ≈70KU/mg
Color Off-White
Inhibitors Heavy Metal ions and is competitively inhibited by DNA
Source Bovine pancreas
pH 7 to 7.5
Molecular Weight ≈13700
Quantity 5g
For Use With (Application) purification of DNA, analysis of RNA sequences, RNase protection assays, hydrolyze RNA contained in Protein samples
Storage Requirements 0°C