Please login to your online account to display your discounted pricing

MP Biomedicals Ribonuclease From Bovine Pancreas

$511.37 - $1,927.58

Specifications

Color Off-White
Product Name Ribonuclease
For Use With (Application) purification of DNA, analysis of RNA sequences, RNase protection assays, hydrolyze RNA contained in Protein samples
Inhibitors Heavy Metal ions and is competitively inhibited by DNA
Molecular Weight ≈13700
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10107505

 
mp biomedicals
0210107505
5g Each for $1,927.58

ICN10107501

 
mp biomedicals
0210107501
1g Each for $511.37
Description & Specifications

Specifications

Color Off-White
Product Name Ribonuclease
For Use With (Application) purification of DNA, analysis of RNA sequences, RNase protection assays, hydrolyze RNA contained in Protein samples
Inhibitors Heavy Metal ions and is competitively inhibited by DNA
Molecular Weight ≈13700
pH 7 to 7.5
Source Bovine pancreas
Storage Requirements 0°C