missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ RapidPURE™ Plasmid Mini Kit

For use in routine molecular biology applications such as fluorescent and radioactive sequencing and cloning

$216.80 - $964.51

Specifications

For Use With (Application) Isolate up to 20ug of plasmid DNA from 1.5mL of an overnight bacterial culture
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
MP112066200 View Documents MP Biomedicals™
112066200
Each for $216.80
 
MP112066400 View Documents MP Biomedicals™
112066400
Each for $406.49
 
MP112066600 View Documents MP Biomedicals™
112066600
Each for $964.51
 
Description

Description

  • The Kit is designed to isolate up to 20μg of plasmid DNA from 1.5mL of an overnight bacterial culture.
  • All steps can be carried out in either a microcentrifuge or vacuum manifold.
Specifications

Specifications

Isolate up to 20ug of plasmid DNA from 1.5mL of an overnight bacterial culture