Please login to your online account to display your discounted pricing

MP Biomedicals Pyruvate Oxidase

Manufacturer: mp biomedicals  0215348680

 View all options for this product

Catalog No. ICN15348680


Description & Specifications


Product Name Pyruvate Oxidase
Format Lyophilized Powder
Concentration ≥1.5μ/mg
Color Yellow
Inhibitors Fe++, Zn++, Cu++, Ag+, Hg++
Source Microorganism
pH 5.7
Quantity 100U
For Use With (Application) Study of Pyruvate metabolism determination of Pyruvate, GOT and GPT
Storage Requirements -20°C