Please login to your online account to display your discounted pricing

MP Biomedicals Pyruvate Oxidase

$441.70 - $441.70

Specifications

Color Yellow
Product Name Pyruvate Oxidase
For Use With (Application) Study of Pyruvate metabolism determination of Pyruvate, GOT and GPT
Inhibitors Fe++, Zn++, Cu++, Ag+, Hg++
pH 5.7
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15348680

 
mp biomedicals
0215348680
100U Each for $441.70
Description & Specifications

Specifications

Color Yellow
Product Name Pyruvate Oxidase
For Use With (Application) Study of Pyruvate metabolism determination of Pyruvate, GOT and GPT
Inhibitors Fe++, Zn++, Cu++, Ag+, Hg++
pH 5.7
Source Microorganism
Storage Requirements -20°C