missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Pyruvate Kinase from Rabbit Muscle
≥200u/mg protein, White Crystalline Powder
Manufacturer: MP Biomedicals™ 0215199925
Specifications
Pyruvate Kinase | |
Lyophilized Powder | |
Clear | |
7.5 | |
25,000U | |
-20°C |
Rabbit muscle | |
≥200U/mg | |
Activity is inhibited by Ca2+, Fluorophosphate inhibits the Enzyme reaction competitively with respect to PEP , ATP inhibits the reaction by removal of Mg2+ from the Substrate MgADP- | |
237000 | |
Used as a coupling Enzyme in conjunction with Lactic Dehydrogenase in quantifying ADP and the activity of Enzymes that catalyze the formation of ADP |