missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Peroxidase from Horseradish Roots
Ideal for production of peroxidase conjugated antisera
$115.52 - $2,518.94
Specifications
Color | Brown to Red Brown |
---|---|
For Use With (Application) | Production of Peroxidase Conjugated Antisera, ELISA, immunohistochemistry, western blotting |
Content And Storage | -20°C |
Source | Horseradish roots |
Product Type | Peroxidase |
Description
- From Horseradish Roots
- Activity: ≥200 units/mg
Specifications
Brown to Red Brown | |
-20°C | |
Peroxidase | |
Lyophilized Powder |
Production of Peroxidase Conjugated Antisera, ELISA, immunohistochemistry, western blotting | |
Horseradish roots | |
Sodium Azide, Cyanide, L-cystine, Dichromate, Ethylenethiourea, Hydroxylamine, Sulfide, Vanadate, p-aminobenzoic Acid and Cd2+, Co2+, Cu2+, Fe3+, Mn2+, Ni2+ and Pb2+ ions |