MP Biomedicals™ NZYM Broth
For the growth of lambda phage in E. coli in liquid medium
Manufacturer:  MP Biomedicals™ 113005022
 View more versions of this product
Catalog No. MP113005022
Specifications
Specifications
Molecular biology Certified™
NZ Amine 10g/L, Yeast extract 5g/L, MgSO4.H2O 2g/L, NaCl 5g/L
15-30°C
For growth of lambda phage in Escherichia coli in liquid medium
Powder
454g
NZYM Broth
Documents