Please login to your online account to display your discounted pricing

MP Biomedicals Molecular Genetics Agar


Suitable as a component in culture media for molecular genetics

Manufacturer: mp biomedicals  19402291

 View more versions of this product

Catalog No. ICN19402291

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 15-30°C
Product Type Agar
Quantity 1000g
Format Powder
For Use With (Application) Suitable as a component in culture media for molecular Genetics
Grade Molecular biology