missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Lysing Matrix S, 2mL tubes

Use to break hard or brittle samples with minimal amounts of mechanical shearing. Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.

Manufacturer:  MP Biomedicals™ 116925100Q#7513

 View more versions of this product

Catalog No. MP116925100



Description

Description

  • Matrix Type: S
  • Tube Size: 2mL
  • Sample Type: All samples, tough tissues, skeletal muscle, lung, heart, bone, seeds, spores
  • Molecule to be Enriched: DNA, RNA, proteins, other small molecules
  • Matrix Composition: Matrix S contains stainless steel
Specifications

Specifications

Dnase-free
Lysing matrix S
RNase-free
Nucleic acid isolation and purification
100 x 2mL Tubes
Room Temperature
Documents