missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Lysing Matrix Y

For lysis of gram positive and negative bacteria, fungal tissue and spores, yeasts, molds, filamentous algaes and biofilms

Manufacturer:  MP Biomedicals™ 116975025

 View more versions of this product

Catalog No. MP116975025



Description

Description

  • Impact-resistant 2mL tube contains 0.5mm diameter Yttria-Stabilized Zirconium Oxide beads
  • Specific garvity: 6.02

Applications:

Nucleic Acid Isolation and Purification

Specifications

Specifications

For cracking tough cell walls For cracking tough cell walls
Lysing matrix Y
25 x 15mL Tubes
Documents