missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Lysing Matrix M

Tubes for optimal lysis used with FastPrep™ instruments

Manufacturer:  MP Biomedicals™ 116923500

 View more versions of this product

Catalog No. MP116923500



Description

Description

  • Impact-resistant 2mL tube contains one 0.24 in. (6.35 mm) diameter Zirconium Oxide coated ceramic grinding sphere
  • Specific gravity: 5.5 to 6.1

Nucleic Acid Isolation and Purification

Specifications

Specifications

Dnase-free
Extra bag of spheres
500 x 2mL Tubes
Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis
Lysing matrix M
RNase-free
Documents