missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Lysing Matrix I
Lysing Matrix I is effective at breaking very tough and hard samples, including organisms with a tough or dense exterior matrix.
$254.38 - $498.50
Specifications
Sample Type | Seeds, Spores, Animal Tissues, Plants, Soil, Ancient and Dried Samples, Clay, Wood |
---|---|
DNase | Dnase-free |
For Use With (Application) | Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis |
Matrix | Lysing matrix I |
RNase | RNase-free |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
MP116918050
|
MP Biomedicals™
116918050 |
50 x 2 mL Tubes |
Each for $254.38
|
|
|||||
MP116918100
|
MP Biomedicals™
116918100 |
100 x 2 mL Tubes |
Each for $498.50
|
|
|||||
Description
- Tube Size: 2 mL
- Sample Type: Plant and animal tissues, wood, seeds, very dense soils and clays, whole insects, ticks, ancient and dried samples, dry grinding fungal spores/rusts
- Molecule to be Enriched: DNA, RNA, proteins, other small molecules
- Matrix Composition: Matrix I, contains 2 mm yellow zirconia beads and a 4 mm black ceramic bead
Specifications
Seeds, Spores, Animal Tissues, Plants, Soil, Ancient and Dried Samples, Clay, Wood | |
Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis | |
RNase-free |
Dnase-free | |
Lysing matrix I |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.