missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Lysing Matrix H

Used to isolate RNA and Proteins from bacterial aggregates, biofilms, and whole insects

Manufacturer:  MP Biomedicals™ 116917050

 View more versions of this product

Catalog No. MP116917050



Description

Description

  • Impact-resistant 2mL tube contains 2mm glass beads and 2mm yellow zirconium oxide beads
  • With yellow caps
  • Specific Gravity: Glass Beads, 2mm; Yellow Zirconium Oxide Beads, 5.5

Applications:

Nucleic Acid Isolation and Purification

Specifications

Specifications

Dnase-free
Lysing matrix H
RNase-free
Nucleic Acid isolation and purification
50 x 2mL Tubes
Documents