missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Lysing Matrix H

Used to isolate RNA and Proteins from bacterial aggregates, biofilms, and whole insects

$262.01 - $2,862.73

Specifications

Matrix Lysing matrix H
View More Specs

Products 3
Catalog Number Mfr. No. For Use With (Application) Quantity Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Quantity Price Quantity  
MP116917050 View Documents MP Biomedicals™
116917050
Nucleic Acid isolation and purification 50 x 2mL Tubes Each for $262.01
 
116917500 View Documents MP Biomedicals™
116917500
Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis 500 x 2mL Tubes Pack for $2,862.73
 
MP116917100 View Documents MP Biomedicals™
116917100
Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis 100 x 2mL Tubes Each for $611.45
 
Description

Description

  • Impact-resistant 2mL tube contains 2mm glass beads and 2mm yellow zirconium oxide beads
  • With yellow caps
  • Specific Gravity: Glass Beads, 2mm; Yellow Zirconium Oxide Beads, 5.5

Applications:

Nucleic Acid Isolation and Purification

Specifications

Specifications

Lysing matrix H