missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Lysing Matrix C, 2mL tubes

Contains 1mm silica spheres. Lysing Matrix C is used primarily for lysis of yeast and fungi and can also be used for molds, spores and filamentous algae.

Manufacturer:  MP Biomedicals™ 116912500

 View more versions of this product

Catalog No. MP116912500



Description

Description

  • Tube Size: 2mL
  • Sample Type:Yeast and yeast spores, fungi and fungal spores, algae
  • Molecule to be Enriched: DNA, RNA, Proteins, other small molecules
Specifications

Specifications

Dnase-free
Lysing matrix C
RNase-free
Nucleic Acid isolation and purification
500 x 2mL Tubes
Room Temperature
Documents
Promotions

Promotions