missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Lysing Matrix B, 96-well racks (1.2mL)

Contains 0.1mm silica spheres. Used primarily for lysis of gram positive and gram negative bacteria, and will also disrupt fungal tissue and spores. Lysing Matrix B can also be used as an additive for disruption by sonication.

Manufacturer:  MP Biomedicals™ 116981010

 View more versions of this product

Catalog No. MP116981010



Description

Description

    Sample Type: Bacteria, Spores
  • Molecule to be Enriched: DNA, RNA, Proteins, other small molecules
Specifications

Specifications

Dnase-free
1.2mL Cluster-strip tube and cap assemblies
10 x 96-Well Plate
Room Temperature
Nucleic acid isolation and purification
Lysing matrix B
RNase-free
Documents