missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Lysing Matrix A, 2mL tubes

For all sample types except soil for the subsequent isolation of genomic DNA. MP Biomedicals Lysing Matrix A impact-resistant tubes contains garnet matrix and one 1/4 in. ceramic sphere.

Manufacturer:  MP Biomedicals™ 116910050*

 View more versions of this product

Catalog No. MP116910050



Description

Description

  • Tube Size: 2mL
  • Sample Type: All Samples
  • Molecule to be Enriched: DNA
  • Grinding matrix is chemically inert and will not bind nucleic acids
Specifications

Specifications

Dnase-free
Lysing matrix A
RNase-free
Nucleic Acid isolation and purification
50 x 2mL Tubes
Room Temperature
Documents
Promotions

Promotions