Please login to your online account to display your discounted pricing

MP Biomedicals Linbro™/Titertek™ EIA Plates


Manufacturer: mp biomedicals  7618104

 View more versions of this product

Catalog No. ICN7618104

  • / Case
 

Description & Specifications

Specifications

Material Polystyrene
Sterile No
Volume (Metric) Well 0.35mL
Type EIA II Microplate
Color Clear