Please login to your online account to display your discounted pricing

MP Biomedicals Linbro™/Titertek™ EIA Plates

$287.88 - $287.88

Specifications

Color Clear
Material Polystyrene
Sterile No
Volume (Metric) Well 0.35mL
View More Specs
Products
Catalog Number Mfr. No. Type Price Quantity    

ICN7618104

 
mp biomedicals
7618104
EIA II Microplate Case for $287.88
Description & Specifications

Specifications

Color Clear
Material Polystyrene
Sterile No
Volume (Metric) Well 0.35mL