Please login to your online account to display your discounted pricing

MP Biomedicals Linbro™ Tissue Culture Plates


Manufacturer: mp biomedicals  7605305

 View more versions of this product

Catalog No. ICN7605305

  • / Each
 

Description & Specifications

Specifications

Lid Yes
Volume (Metric) 7.5mL
Format 12-well
Material Polystyrene
No. of Wells 12
Width (Metric) Exterior 8.4cm
Closure Yes
Color Clear
Sterile Yes
Type Tissue Culture Plate
For Use With (Application) Ideal for a variety of applications including cell cloning and metabolic labeling procedures
Culture Area 4.5cm2
Length (Metric) Exterior 11.1cm