missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Linbro™ Microplate Sealer

For use with multi-well plates and tissue culture plates

$58.16 - $564.14

Specifications

Material Acetate
Type Microplate Sealer
Temperature Range (Metric) -16°C to +38°C
Color Clear
For Use With (Application) Used for multi-well plates and tissue culture plates
View More Specs

Products 2
Catalog Number Mfr. No. Material No. per Pack Price Quantity  
Catalog Number Mfr. No. Material No. per Pack Price Quantity  
ICN7640105
View Documents
MP Biomedicals™
097640105
Acetate 100 Each for $58.16
 
MP0976401C2
View Documents
MP Biomedicals™
0976401C2
Acetate 1000 Pack of 1000 Each for $564.14
 
Description

Description

  • Easy application and removal for all microplates for both short- and long-term storage Provide tight seal to minimize evaporation and condensation
  • Suitable for use between -16 °C and 38 °C
  • Transparent tape and is not pierceable
  • Nonsterile
Specifications

Specifications

Acetate
-16°C to +38°C
Used for multi-well plates and tissue culture plates
Microplate Sealer
Clear
Non-sterile