missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Linbro™ Microplate Sealer

For use with multi-well plates and tissue culture plates

Manufacturer:  MP Biomedicals™ 097640105

 View more versions of this product

Catalog No. ICN7640105



Description

Description

  • Easy application and removal for all microplates for both short- and long-term storage Provide tight seal to minimize evaporation and condensation
  • Suitable for use between -16 °C and 38 °C
  • Transparent tape and is not pierceable
  • Nonsterile
Specifications

Specifications

Used for multi-well plates and tissue culture plates
-16°C to +38°C
Acetate
Microplate Sealer
100
Clear
Non-sterile
Documents