Please login to your online account to display your discounted pricing

MP Biomedicals Linbro™ 2.0mL Snap-Top Microcentrifuge Tubes


With reference lines at 0.5, 1.0, 1.5, and 2.0mL for quick volume estimations

Manufacturer: mp biomedicals  0219352390

 View more versions of this product

Catalog No. ICN19352390

  • / Pack of 500 Each
 

Description & Specifications

Specifications

Cap Style Snap Cap
Closure Type Snap
Capacity (Metric) 2mL
Color Blue
Material Plastic
For Use With (Application) For those experiments that entail large numbers of samples that may be difficult to organize and keep track of
Graduations 0.5, 1, 1.5, and 2mL
Packaging 500/Cs.
Product Type Microcentrifuge Tube

Frosted writing area.