MP Biomedicals™ Linbro™ 2.0mL Snap-Top Microcentrifuge Tubes
With reference lines at 0.5, 1.0, 1.5, and 2.0mL for quick volume estimations
Manufacturer:  MP Biomedicals™ 19353090
 View more versions of this product
Catalog No. ICN19353090
Description
Description
Frosted writing area.
Specifications
Specifications
Snap Cap
Yes
Amber
For those experiments that entail large numbers of samples that may be difficult to organize and keep track of
500/Cs.
Snap
2mL
Plastic
0.5, 1, 1.5, and 2mL
Documents