missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ LB Medium, Dehydrated: Bulk Powder

Formulations of the E. coli growth media known as LB Medium typically contain 10g of tryptone and 5g of yeast extract per liter; contains 10g NaCl per liter

$120.84 - $921.18

Specifications

Certifications/Compliance Molecular Biology Certified™
Format Dehydrated
For Use With (Application) For growth of bacteria
Packaging Bulk Powder
Grade Molecular Biology
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MP113002022 View Documents MP Biomedicals™
113002022
454g Each for $120.84
MP113002032 View Documents MP Biomedicals™
113002032
1000g Each for $232.63
MP113002042 View Documents MP Biomedicals™
113002042
5000g Each for $921.18
Specifications

Specifications

Molecular Biology Certified™
For growth of bacteria
Molecular Biology
15-30°C
Dehydrated
Bulk Powder
Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L