missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ LB Medium, Dehydrated: Bulk Powder

Formulations of the E. coli growth media known as LB Medium typically contain 10g of tryptone and 5g of yeast extract per liter; contains 10g NaCl per liter

Manufacturer:  MP Biomedicals™ 113002042

 View more versions of this product

Catalog No. MP113002042



Specifications

Specifications

Molecular Biology Certified™
Dehydrated
Bulk Powder
15-30°C
For growth of bacteria
Molecular Biology
Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L
5000g
LB medium
Documents