Please login to your online account to display your discounted pricing

MP Biomedicals L-Amino Acid Oxidase, Crotalus adamanteus from Crotalus adamanteus

Used to convert L-amino acids to their corresponding α-keto acids

Manufacturer: mp biomedicals  0210027210

 View all options for this product

Catalog No. ICN10027210


Description & Specifications


Product Name L-Amino Acid Oxidase
Format Solution
Concentration ≥4U/mg
Color Clear, Yellow
Source Crotalus adamanteus
Quantity 10mg
For Use With (Application) L-Amino Acid Oxidase is used to convert L-Amino Acids to their corresponding alpha-keto Acids
Storage Requirements 4°C

L-amino acid oxidase is involved in various metabolic pathways such as alanine and aspartate metabolism, methionine metabolism, valine, leucine and isoleucine degradation, tyrosine metabolism, phenylalanine metabolism, tryptophan metabolism, phenylalanine, tyrosine and tryptophan biosynthesis, and alkaloid biosynthesis. It is found in microorganisms and in animal tissue in kidney and liver.

  • CAS Number: 9000-89-9
  • EC Number: 232-564-0
  • MDL Number: MFCD00131454
  • Format: Clear, yellow solution
  • Specific Activity: ≥4U/mg protein