Please login to your online account to display your discounted pricing

MP Biomedicals L-Amino Acid Oxidase, Crotalus adamanteus from Crotalus adamanteus

Used to convert L-amino acids to their corresponding α-keto acids

$192.34 - $719.12

Specifications

Color Clear, Yellow
Product Name L-Amino Acid Oxidase
For Use With (Application) L-Amino Acid Oxidase is used to convert L-Amino Acids to their corresponding alpha-keto Acids
Source Crotalus adamanteus
Storage Requirements 4°C
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10027202

 
mp biomedicals
0210027202
2mg Each for $192.34

ICN10027205

 
mp biomedicals
0210027205
5mg Each for $388.29

ICN10027210

 
mp biomedicals
0210027210
10mg Each for $719.12
Description & Specifications

Specifications

Color Clear, Yellow
Product Name L-Amino Acid Oxidase
For Use With (Application) L-Amino Acid Oxidase is used to convert L-Amino Acids to their corresponding alpha-keto Acids
Source Crotalus adamanteus
Storage Requirements 4°C

L-amino acid oxidase is involved in various metabolic pathways such as alanine and aspartate metabolism, methionine metabolism, valine, leucine and isoleucine degradation, tyrosine metabolism, phenylalanine metabolism, tryptophan metabolism, phenylalanine, tyrosine and tryptophan biosynthesis, and alkaloid biosynthesis. It is found in microorganisms and in animal tissue in kidney and liver.

  • CAS Number: 9000-89-9
  • EC Number: 232-564-0
  • MDL Number: MFCD00131454
  • Format: Clear, yellow solution
  • Specific Activity: ≥4U/mg protein