missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals Hyaluronidase, Source: Ovine Testes

Ovine Testes

$529.20 - $529.20

Specifications

Color White
Product Name Hyaluronidase
For Use With (Application) Drug dispersion and delivery
Inhibitors Fe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+
Molecular Weight ≈55
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15127201 MP Biomedicals
15127201
1g Each for $529.20
Specifications

Description

Hyaluronidase, [37326-33-3]

  • Hyaluronate Glycanohydrolase
  • Lyophilized, Salt-free
  • Activity: 500 I.U./mg material

Description & Specifications

Specifications

White
Drug dispersion and delivery
≈55
Ovine testes
Hyaluronidase
Fe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+
4.5 to 6
-20°C