Please login to your online account to display your discounted pricing

MP Biomedicals Hyaluronidase, Ovine testes


Source: Ovine Testes

Manufacturer: mp biomedicals  15127580

 View more versions of this product

Catalog No. ICN15127580

  • / Each
 

Description & Specifications

Specifications

pH 4.5 to 6
Concentration >1000U/mg
Color White
Inhibitors Fe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+
Product Name Hyaluronidase
Molecular Weight ≈55
Storage Requirements -20°C
Source Ovine testes
For Use With (Application) Drug dispersion and delivery
Format Lyophilized
Quantity 100mg